Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cuta
Entrez Gene ID
294288
Gene Name
cutA divalent cation tolerance homolog (E. coli)
Gene Aliases
CutA1
Chromosome Location
Chr.20: 5532024 - 5533632 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256120.2 XP_006256182.1 1 - 2 112 126

View Full Product Details