Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf8
Entrez Gene ID
361815
Gene Name
ring finger protein 8, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.20: 8285338 - 8309858 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025727.1 NP_001020898.1 7 - 8 1528 104
GenBank mRNA BC099079.1 - 7 - 8 1528 104

View Full Product Details