The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025750.1 | NP_001020921.1 | 9 - 10 | 1259 | 89 |
GenBank mRNA | BC098846.1 | - | 9 - 10 | 1259 | 89 |
FQ225547.1 | - | 9 - 10 | 1136 | 89 | |
FQ232611.1 | - | 9 - 10 | 1120 | 89 |