The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081972.1 | NP_001075441.1 | 12 - 13 | 1596 | 78 |
NM_001081973.1 | NP_001075442.1 | 12 - 13 | 1619 | 78 | |
NM_001081974.1 | NP_001075443.1 | 12 - 13 | 1605 | 78 | |
XM_006240621.2 | XP_006240683.1 | 12 - 13 | 2189 | 78 | |
XM_006240623.2 | XP_006240685.1 | 12 - 13 | 2190 | 78 | |
XM_006240625.2 | XP_006240687.1 | 12 - 13 | 2085 | 78 | |
XM_006240626.2 | XP_006240688.1 | 12 - 13 | 2084 | 78 | |
XM_006240627.2 | XP_006240689.1 | 12 - 13 | 2085 | 78 | |
XM_006240629.2 | XP_006240691.1 | 12 - 13 | 2084 | 78 | |
GenBank mRNA | M75146.1 | - | 12 - 13 | 1596 | 78 |
M75147.1 | - | 12 - 13 | 1619 | 78 | |
M75148.1 | - | 12 - 13 | 1605 | 78 |