Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd6
Entrez Gene ID
690367
Gene Name
BTB (POZ) domain containing 6
Gene Aliases
-
Chromosome Location
Chr.6: 137785665 - 137788442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077683.2 NP_001071151.2 1 - 2 470 136
XM_006240664.2 XP_006240726.1 2 - 3 624 136
GenBank mRNA DV213761.1 - 1 - 2 470 136

View Full Product Details