The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013170.2 | NP_001013188.2 | 10 - 11 | 1389 | 106 |
XM_006240542.2 | XP_006240604.1 | 10 - 11 | 1426 | 106 | |
XM_008764811.1 | XP_008763033.1 | 10 - 11 | 1580 | 106 | |
XM_008764812.1 | XP_008763034.1 | 10 - 11 | 2177 | 106 | |
GenBank mRNA | AY724504.1 | - | 10 - 11 | 1389 | 106 |