The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025723.1 | NP_001020894.1 | 6 - 7 | 985 | 154 |
GenBank mRNA | BC097970.1 | - | 6 - 7 | 985 | 154 |