Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnksr3
Entrez Gene ID
308113
Gene Name
Cnksr family member 3
Gene Aliases
Magi1, Prp4
Chromosome Location
Chr.1: 43789797 - 43884267 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012061.1 NP_001012061.1 7 - 8 1249 71
XM_006227875.2 XP_006227937.1 7 - 8 2406 71
GenBank mRNA AY328890.1 - 7 - 8 1249 71

View Full Product Details