The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006982.1 | NP_001006983.1 | 5 - 6 | 420 | 83 |
XM_006256611.2 | XP_006256673.1 | 6 - 7 | 477 | 83 | |
GenBank mRNA | BC082752.1 | - | 5 - 6 | 420 | 83 |
FQ210799.1 | - | 6 - 7 | 465 | 83 | |
FQ221998.1 | - | 6 - 7 | 468 | 83 |