Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otc
Entrez Gene ID
25611
Gene Name
ornithine carbamoyltransferase
Gene Aliases
-
Chromosome Location
Chr.X: 13524804 - 13601074 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013078.1 NP_037210.1 2 - 3 315 123
GenBank mRNA BC088158.1 - 2 - 3 281 123
FQ211098.1 - 2 - 3 299 123
FQ219225.1 - 2 - 3 302 123
FQ219657.1 - 2 - 3 278 123
K03040.1 - 2 - 3 273 123
M11266.1 - 2 - 3 315 123
X01178.1 - 2 - 3 315 123

View Full Product Details