Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spaca5
Entrez Gene ID
314431
Gene Name
sperm acrosome associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 912770 - 915953 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108058.1 NP_001101528.1 1 - 2 146 78

View Full Product Details