Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nyx
Entrez Gene ID
302516
Gene Name
nyctalopin
Gene Aliases
RGD1561300
Chromosome Location
Chr.X: 10197547 - 10218583 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100967.1 NP_001094437.1 1 - 2 144 122
GenBank mRNA DQ393414.1 - 1 - 2 144 122

View Full Product Details