The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107433.1 | NP_001100903.1 | 3 - 4 | 1409 | 67 |
XM_006255674.2 | XP_006255736.1 | 5 - 6 | 1930 | 67 | |
XM_006255675.2 | XP_006255737.1 | 5 - 6 | 1744 | 67 | |
XM_008772515.1 | XP_008770737.1 | 4 - 5 | 1959 | 67 |