Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo31
Entrez Gene ID
498959
Gene Name
F-box protein 31
Gene Aliases
RGD1561069
Chromosome Location
Chr.19: 53487616 - 53625673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044259.2 NP_001037724.2 3 - 4 450 77
XM_006255719.2 XP_006255781.1 3 - 4 1444 77
XM_006255720.2 XP_006255782.1 2 - 3 1357 77
GenBank mRNA BC110046.1 - 1 - 2 556 77
BC166840.1 - 3 - 4 450 77

View Full Product Details