The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106194.1 | NP_001099664.1 | 5 - 6 | 672 | 83 |
NM_001201369.1 | NP_001188298.1 | 6 - 7 | 821 | 83 | |
GenBank mRNA | BF283670.1 | - | 1 - 2 | 83 | |
BQ781676.1 | - | 1 - 2 | 83 |