The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022962.1 | NP_075251.1 | 23 - 24 | 3960 | 107 |
GenBank mRNA | AF081144.1 | - | 22 - 23 | 3945 | 107 |
AF081145.1 | - | 21 - 22 | 4077 | 107 | |
AF081146.1 | - | 23 - 24 | 3960 | 107 | |
AF081147.1 | - | 22 - 23 | 4092 | 107 | |
AF111099.1 | - | 24 - 25 | 4286 | 107 | |
U72487.1 | - | 22 - 23 | 3762 | 107 | |
U78105.1 | - | 22 - 23 | 4087 | 107 |