Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lphn1
Entrez Gene ID
65096
Gene Name
latrophilin 1
Gene Aliases
CL1BA
Chromosome Location
Chr.19: 25029037 - 25069146 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022962.1 NP_075251.1 8 - 9 1908 87
GenBank mRNA AF081144.1 - 7 - 8 1893 87
AF081145.1 - 7 - 8 1893 87
AF081146.1 - 8 - 9 1908 87
AF081147.1 - 8 - 9 1908 87
AF111099.1 - 9 - 10 2234 87
U72487.1 - 7 - 8 1710 87
U78105.1 - 7 - 8 2035 87

View Full Product Details