Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsd11b2
Entrez Gene ID
25117
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.19: 37476083 - 37481326 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017081.2 NP_058777.1 3 - 4 793 137
XM_006255445.2 XP_006255507.1 3 - 4 1509 137
GenBank mRNA BC087023.1 - 3 - 4 780 137
U22424.1 - 3 - 4 794 137

View Full Product Details