The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008772666.1 | XP_008770888.1 | 2 - 3 | 207 | 85 |
XM_008772667.1 | XP_008770889.1 | 2 - 3 | 207 | 85 | |
XM_008772668.1 | XP_008770890.1 | 1 - 2 | 101 | 85 |