Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkn1
Entrez Gene ID
29355
Gene Name
protein kinase N1
Gene Aliases
PAK-1, Prkcl1
Chromosome Location
Chr.19: 24803524 - 24831500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017175.2 NP_058871.2 11 - 12 1817 84
XM_006255245.2 XP_006255307.1 11 - 12 2857 84
GenBank mRNA BC061836.1 - 11 - 12 1718 84
D26180.1 - 11 - 12 1768 84
FQ222217.1 - 11 - 12 1817 84
L35634.1 - 11 - 12 1660 84

View Full Product Details