Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Calb2
Entrez Gene ID
117059
Gene Name
calbindin 2
Gene Aliases
-
Chromosome Location
Chr.19: 41475795 - 41509630 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053988.1 NP_446440.1 3 - 4 315 113
XM_008772466.1 XP_008770688.1 4 - 5 494 113
GenBank mRNA BC087603.1 - 3 - 4 285 113
X66974.1 - 3 - 4 315 113

View Full Product Details