Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adat1
Entrez Gene ID
690810
Gene Name
adenosine deaminase, tRNA-specific 1
Gene Aliases
-
Chromosome Location
Chr.19: 44175865 - 44212101 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109611.1 NP_001103081.1 8 - 9 1428 116
XM_006255634.2 XP_006255696.1 8 - 9 2268 116
XM_006255635.2 XP_006255697.1 8 - 9 2394 116
XM_006255636.2 XP_006255698.1 8 - 9 2222 116
XM_006255638.1 XP_006255700.1 7 - 8 1273 116
XM_008772535.1 XP_008770757.1 8 - 9 2268 116

View Full Product Details