The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100505.1 | NP_001093975.1 | 5 - 6 | 726 | 61 |
XM_008772367.1 | XP_008770589.1 | 6 - 7 | 1547 | 61 | |
XM_008772368.1 | XP_008770590.1 | 6 - 7 | 1675 | 61 | |
XM_008772369.1 | XP_008770591.1 | 6 - 7 | 1670 | 61 | |
GenBank mRNA | BC098819.1 | - | 5 - 6 | 661 | 61 |