Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cenpn
Entrez Gene ID
361416
Gene Name
centromere protein N
Gene Aliases
Da2-27, Da2-4, RGD1310953
Chromosome Location
Chr.19: 49457462 - 49479996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008366.1 NP_001008367.1 10 - 11 1017 89
XM_006255686.2 XP_006255748.1 10 - 11 1131 89
XM_006255687.2 XP_006255749.1 10 - 11 1049 89
XM_006255689.2 XP_006255751.1 11 - 12 1353 89
XM_006255690.2 XP_006255752.1 11 - 12 1139 89
XM_006255691.2 XP_006255753.1 9 - 10 1073 89
XM_006255692.2 XP_006255754.1 9 - 10 953 89
GenBank mRNA AY325257.1 - - 1304 89
AY325261.1 - - 1304 89
BC085839.1 - 10 - 11 1017 89

View Full Product Details