Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cenpn
Entrez Gene ID
361416
Gene Name
centromere protein N
Gene Aliases
Da2-27, Da2-4, RGD1310953
Chromosome Location
Chr.19: 49457462 - 49479996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008366.1 NP_001008367.1 5 - 6 494 141
XM_006255686.2 XP_006255748.1 5 - 6 608 141
XM_006255687.2 XP_006255749.1 5 - 6 526 141
XM_006255689.2 XP_006255751.1 6 - 7 830 141
XM_006255690.2 XP_006255752.1 6 - 7 616 141
GenBank mRNA BC085839.1 - 5 - 6 494 141

View Full Product Details