Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txnl4b
Entrez Gene ID
292008
Gene Name
thioredoxin-like 4B
Gene Aliases
RGD1305127
Chromosome Location
Chr.19: 42101081 - 42110222 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255579.2 XP_006255641.1 1 - 2 99 95

View Full Product Details