Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rltpr
Entrez Gene ID
307797
Gene Name
RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing
Gene Aliases
RGD1562390
Chromosome Location
Chr.19: 37652384 - 37665333 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255548.2 XP_006255610.1 34 - 35 4335 110
XM_008772559.1 XP_008770781.1 34 - 35 4335 110

View Full Product Details