The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004277.2 | NP_001004277.1 | 5 - 6 | 762 | 109 |
GenBank mRNA | AY490816.1 | - | 5 - 6 | 734 | 109 |
BC098894.1 | - | 5 - 6 | 762 | 109 |