Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acd
Entrez Gene ID
307798
Gene Name
adrenocortical dysplasia homolog (mouse)
Gene Aliases
-
Chromosome Location
Chr.19: 37665295 - 37668041 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037193.1 NP_001032270.1 6 - 7 526 72
XM_008772496.1 XP_008770718.1 6 - 7 580 72
GenBank mRNA BC099222.1 - 6 - 7 526 72

View Full Product Details