Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Marveld3
Entrez Gene ID
498950
Gene Name
MARVEL domain containing 3
Gene Aliases
RGD1562056
Chromosome Location
Chr.19: 41710076 - 41724923 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255598.2 XP_006255660.1 2 - 3 647 67

View Full Product Details