Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Palm3
Entrez Gene ID
100359592
Gene Name
paralemmin 3
Gene Aliases
-
Chromosome Location
Chr.19: 25143488 - 25151940 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728582.4 XP_002728628.1 3 - 4 281 73

View Full Product Details