Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcng4
Entrez Gene ID
307900
Gene Name
potassium channel, voltage gated modifier subfamily G, member 4
Gene Aliases
-
Chromosome Location
Chr.19: 52270907 - 52283229 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107435.1 NP_001100905.1 2 - 3 1241 73
XM_006255709.2 XP_006255771.1 3 - 4 1809 73
XM_006255710.2 XP_006255772.1 3 - 4 1813 73
XM_006255711.2 XP_006255773.1 3 - 4 1455 73
XM_008772608.1 XP_008770830.1 2 - 3 1239 73

View Full Product Details