Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Terf2
Entrez Gene ID
361403
Gene Name
telomeric repeat binding factor 2
Gene Aliases
-
Chromosome Location
Chr.19: 39271992 - 39301500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108448.1 NP_001101918.1 7 - 8 1245 76

View Full Product Details