Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfpm1
Entrez Gene ID
691504
Gene Name
zinc finger protein, multitype 1
Gene Aliases
-
Chromosome Location
Chr.19: 55094585 - 55150994 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242627.1 NP_001229556.1 1 - 2 74 62

View Full Product Details