Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd97
Entrez Gene ID
361383
Gene Name
CD97 molecule
Gene Aliases
-
Chromosome Location
Chr.19: 24854981 - 24875712 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012164.1 NP_001012164.1 19 - 20 2481 95
XM_006255278.2 XP_006255340.1 19 - 20 3148 95
XM_006255279.2 XP_006255341.1 18 - 19 3013 95
XM_006255280.1 XP_006255342.1 20 - 21 2730 95
XM_006255281.2 XP_006255343.1 17 - 18 2866 95
GenBank mRNA BC083857.1 - 19 - 20 2481 95

View Full Product Details