Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd97
Entrez Gene ID
361383
Gene Name
CD97 molecule
Gene Aliases
-
Chromosome Location
Chr.19: 24854981 - 24875712 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012164.1 NP_001012164.1 2 - 3 106 102
XM_006255278.2 XP_006255340.1 2 - 3 791 102
XM_006255279.2 XP_006255341.1 2 - 3 791 102
XM_006255280.1 XP_006255342.1 3 - 4 373 102
XM_006255281.2 XP_006255343.1 2 - 3 791 102
GenBank mRNA BC083857.1 - 2 - 3 106 102

View Full Product Details