Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inadl
Entrez Gene ID
140581
Gene Name
InaD-like (Drosophila)
Gene Aliases
Inadl2, RGD1565362
Chromosome Location
Chr.5: 117038615 - 117339923 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238423.2 XP_006238485.1 30 - 31 4186 67
XM_006238424.2 XP_006238486.1 30 - 31 4192 67
XM_006238425.2 XP_006238487.1 5 - 6 1228 67
XM_008763916.1 XP_008762138.1 6 - 7 1282 67
GenBank mRNA FQ212190.1 - 6 - 7 903 67

View Full Product Details