Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inadl
Entrez Gene ID
140581
Gene Name
InaD-like (Drosophila)
Gene Aliases
Inadl2, RGD1565362
Chromosome Location
Chr.5: 117038615 - 117339923 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238423.2 XP_006238485.1 7 - 8 1111 146
XM_006238424.2 XP_006238486.1 7 - 8 1117 146
XM_008763915.1 XP_008762137.1 7 - 8 1111 146

View Full Product Details