Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp2j16
- Entrez Gene ID
-
502969
- Gene Name
- cytochrome P450, family 2, subfamily j, polypeptide 16
- Gene Aliases
-
RGD1561242
- Chromosome Location
-
Chr.5: 119455819 - 119505395 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_578474.1 |
XP_578474.1 |
4 - 5 |
696 |
91 |
View Full Product Details