Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ascc1
Entrez Gene ID
294512
Gene Name
activating signal cointegrator 1 complex subunit 1
Gene Aliases
Asc-1, CGI-18
Chromosome Location
Chr.20: 29558330 - 29648899 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007632.2 NP_001007633.1 5 - 6 556 95
XM_006256404.2 XP_006256466.1 5 - 6 750 95
XM_006256405.2 XP_006256467.1 5 - 6 747 95
XM_006256407.2 XP_006256469.2 5 - 6 636 95
XM_006256408.2 XP_006256470.1 5 - 6 705 95
XM_006256409.1 XP_006256471.1 4 - 5 491 95
XM_006256410.2 XP_006256472.1 5 - 6 702 95
XM_008772881.1 XP_008771103.1 4 - 5 488 95
GenBank mRNA BP504504.1 - 1 - 2 47 95
CK357599.1 - 5 - 6 556 95
FQ226270.1 - 2 - 3 203 95

View Full Product Details