Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps26a
Entrez Gene ID
361846
Gene Name
vacuolar protein sorting 26 homolog A (S. pombe)
Gene Aliases
Vps26
Chromosome Location
Chr.20: 32085465 - 32115310 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007740.1 NP_001007741.1 7 - 8 798 102
XM_006256460.2 XP_006256522.1 7 - 8 921 102
GenBank mRNA BC079150.1 - 7 - 8 798 102
FQ211877.1 - 7 - 8 811 102

View Full Product Details