Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nus1
Entrez Gene ID
294400
Gene Name
nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae)
Gene Aliases
RGD1307879
Chromosome Location
Chr.20: 33557052 - 33584011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164157.1 NP_001157629.1 1 - 2 505 100

View Full Product Details