Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lims1
Entrez Gene ID
499443
Gene Name
LIM and senescent cell antigen-like domains 1
Gene Aliases
Pinch1, RGD1560732
Chromosome Location
Chr.20: 27895983 - 28004767 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145456.1 NP_001138928.1 2 - 3 323 91
XM_006256385.2 XP_006256447.1 2 - 3 2225 91
XM_006256386.2 XP_006256448.1 2 - 3 2225 91
XM_006256387.2 XP_006256449.1 2 - 3 2222 91
XM_006256388.2 XP_006256450.1 2 - 3 2089 91
XM_006256389.2 XP_006256451.1 2 - 3 316 91
XM_006256390.2 XP_006256452.1 2 - 3 1896 91
XM_006256391.2 XP_006256453.1 2 - 3 1895 91
GenBank mRNA FJ640984.1 - 2 - 3 323 91
FJ640985.1 - 2 - 3 402 91
FQ222665.1 - 2 - 3 411 91

View Full Product Details