The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080906.2 | NP_543182.1 | 2 - 3 | 374 | 72 |
GenBank mRNA | AF334162.1 | - | 2 - 3 | 379 | 72 |
BC062021.1 | - | 2 - 3 | 307 | 72 |