Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anapc16
Entrez Gene ID
100360936
Gene Name
anaphase promoting complex subunit 16
Gene Aliases
-
Chromosome Location
Chr.20: 29544626 - 29558928 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256434.1 XP_006256496.1 4 - 5 408 89
XM_008772955.1 XP_008771177.1 3 - 4 353 89
GenBank mRNA FQ209472.1 - 4 - 5 411 89
FQ210195.1 - 3 - 4 356 89

View Full Product Details