The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013907.1 | NP_001013929.1 | 8 - 9 | 1225 | 73 |
GenBank mRNA | BC090076.1 | - | 8 - 9 | 1225 | 73 |