The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135807.1 | NP_001129279.1 | 8 - 9 | 1140 | 76 |
GenBank mRNA | BC161981.1 | - | 8 - 9 | 1140 | 76 |