Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35f1
Entrez Gene ID
502421
Gene Name
solute carrier family 35, member F1
Gene Aliases
RGD1559576
Chromosome Location
Chr.20: 33772314 - 33783387 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109338.1 NP_001102808.1 - 1176 111

View Full Product Details