Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spock2
Entrez Gene ID
361840
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2
Gene Aliases
-
Chromosome Location
Chr.20: 29654579 - 29679927 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108533.2 NP_001102003.1 9 - 10 996 72
XM_006256419.2 XP_006256481.1 9 - 10 1637 72

View Full Product Details