The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014232.1 | NP_001014254.1 | 2 - 3 | 477 | 68 |
GenBank mRNA | AY883936.1 | - | 2 - 3 | 566 | 68 |
BC088754.1 | - | 2 - 3 | 477 | 68 |